Product Details

SNP ID
rs78770668
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:6328698 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCGAGCAAAGAGAGCTCAAAACC[A/C]AACTCACTAAATTGGAGGCTGAAAT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TPD52L3 PubMed Links

Gene Details

Gene
TPD52L3
Gene Name
tumor protein D52-like 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001874.2 350 Missense Mutation AAA,CAA K35Q NP_001001874.1
NM_001001875.3 350 Missense Mutation AAA,CAA K35Q NP_001001875.1
NM_033516.5 350 Missense Mutation AAA,CAA K35Q NP_277051.3
XM_017015280.1 350 Missense Mutation AAA,CAA K35Q XP_016870769.1

View Full Product Details