Product Details

SNP ID
rs75653339
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:109782487 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACATGTACAGACTTTTTCTTGTCA[C/T]CATTCCCTAAATAATACAGTATAAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC107987013 PubMed Links

Gene Details

Gene
LOC107987013
Gene Name
putative uncharacterized protein DDB_G0287265
There are no transcripts associated with this gene.

Gene
PALM2
Gene Name
paralemmin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001037293.2 Intron NP_001032370.1
NM_053016.5 Intron NP_443749.5
Gene
PALM2-AKAP2
Gene Name
PALM2-AKAP2 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007203.4 Intron NP_009134.1
NM_147150.2 Intron NP_671492.1

View Full Product Details