Product Details

SNP ID
rs79598987
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:111611137 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGATTCTCCAGGAAATTTCCTTACT[C/T]GCAAAACTTGATGACAATTTTATCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LRRC37A5P PubMed Links

Gene Details

Gene
LRRC37A5P
Gene Name
leucine rich repeat containing 37 member A5, pseudogene
There are no transcripts associated with this gene.

View Full Product Details