Product Details

SNP ID
rs75464924
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:112887169 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACTTTCTAAGTCACTCCTATACCC[A/G]TCTCCACTCTCTGTGCCCTGGATGG
Phenotype
MIM: 608956
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC46A2 PubMed Links

Gene Details

Gene
SLC46A2
Gene Name
solute carrier family 46 member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033051.3 Intron NP_149040.3

View Full Product Details