Product Details

SNP ID
rs77484121
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:120856230 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGAGGAAAGTGGGTGGGCTGTGC[A/T]CAGGAGGTGAGGTCCCCCTTAAAAG
Phenotype
MIM: 609740
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
PHF19 PubMed Links

Gene Details

Gene
PHF19
Gene Name
PHD finger protein 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001009936.2 3581 Intron NP_001009936.1
NM_001286840.1 3581 UTR 3 NP_001273769.1
NM_001286842.1 3581 UTR 3 NP_001273771.1
NM_001286843.1 3581 Intron NP_001273772.1
NM_015651.2 3581 UTR 3 NP_056466.1
XM_005251906.2 3581 UTR 3 XP_005251963.1
XM_011518509.2 3581 UTR 3 XP_011516811.1
XM_011518511.2 3581 Intron XP_011516813.1
XM_011518515.2 3581 Intron XP_011516817.1
XM_011518516.2 3581 Intron XP_011516818.1
XM_017014612.1 3581 UTR 3 XP_016870101.1
XM_017014613.1 3581 Intron XP_016870102.1
Gene
PSMD5-AS1
Gene Name
PSMD5 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details