Product Details

SNP ID
rs79742783
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:124947257 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTACTCAGTAATAATACTTAAATAT[A/G]GATCAAGAAAATAAAGGACTACTAC
Phenotype
MIM: 602502
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GOLGA1 PubMed Links

Gene Details

Gene
GOLGA1
Gene Name
golgin A1
There are no transcripts associated with this gene.

Gene
SCAI
Gene Name
suppressor of cancer cell invasion
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144877.2 7562 UTR 3 NP_001138349.1
NM_173690.4 7562 UTR 3 NP_775961.2

View Full Product Details