Product Details

SNP ID
rs78878898
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:12131219 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGACTGCCCATCATTTGTAGAGGC[C/T]TTTTTAAAAAAGATAATATAGTCAA
Phenotype
MIM: 614984
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DHTKD1 PubMed Links

Gene Details

Gene
DHTKD1
Gene Name
dehydrogenase E1 and transketolase domain containing 1
There are no transcripts associated with this gene.

Gene
MIR548AK
Gene Name
microRNA 548ak
There are no transcripts associated with this gene.

Gene
SEC61A2
Gene Name
Sec61 translocon alpha 2 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142627.2 Intron NP_001136099.1
NM_001142628.1 Intron NP_001136100.1
NM_018144.3 Intron NP_060614.2

View Full Product Details