Product Details

SNP ID
rs78838870
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:7752572 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCAATTGTGCTCTTTGGTCGTTAC[A/G]TAAATAAGTTGAAAACGTCTATCCA
Phenotype
MIM: 146640 MIM: 601720
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ITIH2 PubMed Links

Gene Details

Gene
ITIH2
Gene Name
inter-alpha-trypsin inhibitor heavy chain 2
There are no transcripts associated with this gene.

Gene
KIN
Gene Name
Kin17 DNA and RNA binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012311.3 4784 UTR 3 NP_036443.1
XM_006717434.3 4784 Intron XP_006717497.1
XM_006717435.3 4784 Intron XP_006717498.1
XM_011519427.1 4784 Intron XP_011517729.1
XM_017015946.1 4784 Intron XP_016871435.1

View Full Product Details