Product Details

SNP ID
rs76840566
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:22828296 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATCTCAGTGTGTAGGAGGGAAGGA[C/T]TGGAGTAATTTCATGAGAGGTATTG
Phenotype
MIM: 602835
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GAS2 PubMed Links

Gene Details

Gene
GAS2
Gene Name
growth arrest specific 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143830.1 Intron NP_001137302.1
NM_005256.3 Intron NP_005247.1
NM_177553.2 Intron NP_808221.1
XM_011519971.2 Intron XP_011518273.1
XM_011519972.2 Intron XP_011518274.1
XM_011519975.1 Intron XP_011518277.1
XM_011519978.2 Intron XP_011518280.1
XM_017017528.1 Intron XP_016873017.1
XM_017017529.1 Intron XP_016873018.1
XM_017017530.1 Intron XP_016873019.1
XM_017017531.1 Intron XP_016873020.1
XM_017017532.1 Intron XP_016873021.1
Gene
SVIP
Gene Name
small VCP interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320340.1 Intron NP_001307269.1
NM_001320341.1 Intron NP_001307270.1
NM_001320342.1 Intron NP_001307271.1
NM_148893.2 Intron NP_683691.1

View Full Product Details