Product Details

SNP ID
rs76455102
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:34105644 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGTCTACCAGTTCCTGAGAGGGA[C/T]GCGTGCCGCGGAGCCAGGCTTACTA
Phenotype
MIM: 601178 MIM: 609221
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CAPRIN1 PubMed Links

Gene Details

Gene
CAPRIN1
Gene Name
cell cycle associated protein 1
There are no transcripts associated with this gene.

Gene
NAT10
Gene Name
N-acetyltransferase 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144030.1 81 UTR 5 NP_001137502.1
NM_024662.2 81 UTR 5 NP_078938.2

View Full Product Details