Product Details

SNP ID
rs78462092
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:4989707 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTCGCCACTCCTTACCCAGGAGAC[A/G]CAAACACAGCTCCTGCAACAATTCC
Phenotype
MIM: 605470
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MMP26 PubMed Links

Gene Details

Gene
MMP26
Gene Name
matrix metallopeptidase 26
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021801.4 180 Silent Mutation ACA,ACG T53T NP_068573.2
XM_011520219.2 180 UTR 5 XP_011518521.1
Gene
OR51L1
Gene Name
olfactory receptor family 51 subfamily L member 1
There are no transcripts associated with this gene.

View Full Product Details