Product Details
- SNP ID
-
rs78676307
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:122278196 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTTCATCCTCGTCTGCCTGAGTGTC[C/T]GTATTACCTTATATTTGAGGAAAAA
- Phenotype
-
MIM: 179838
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CLIP1
PubMed Links
Gene Details
- Gene
- CLIP1
- Gene Name
- CAP-Gly domain containing linker protein 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001247997.1 |
3913 |
Silent Mutation |
ACA,ACG |
T1308T |
NP_001234926.1 |
NM_002956.2 |
3913 |
Silent Mutation |
ACA,ACG |
T1297T |
NP_002947.1 |
NM_198240.1 |
3913 |
Silent Mutation |
ACA,ACG |
T1262T |
NP_937883.1 |
XM_005253593.2 |
3913 |
Silent Mutation |
ACA,ACG |
T1308T |
XP_005253650.1 |
XM_006719552.3 |
3913 |
Silent Mutation |
ACA,ACG |
T1297T |
XP_006719615.1 |
XM_017019786.1 |
3913 |
Silent Mutation |
ACA,ACG |
T1297T |
XP_016875275.1 |
XM_017019787.1 |
3913 |
Silent Mutation |
ACA,ACG |
T1269T |
XP_016875276.1 |
XM_017019788.1 |
3913 |
Silent Mutation |
ACA,ACG |
T1262T |
XP_016875277.1 |
XM_017019789.1 |
3913 |
Silent Mutation |
ACA,ACG |
T1262T |
XP_016875278.1 |
XM_017019790.1 |
3913 |
Silent Mutation |
ACA,ACG |
T1262T |
XP_016875279.1 |
XM_017019791.1 |
3913 |
Silent Mutation |
ACA,ACG |
T1258T |
XP_016875280.1 |
XM_017019792.1 |
3913 |
Silent Mutation |
ACA,ACG |
T1223T |
XP_016875281.1 |
View Full Product Details