Product Details

SNP ID
rs80316885
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:122172552 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCACAGAAATGTTTGTTTCTGGTG[C/T]ATCTTGAAATATGAGTTTGTCGAGG
Phenotype
MIM: 609509
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IL31 PubMed Links

Gene Details

Gene
IL31
Gene Name
interleukin 31
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001014336.1 382 Missense Mutation ACA,GCA T119A NP_001014358.1
Gene
LRRC43
Gene Name
leucine rich repeat containing 43
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098519.1 382 Intron NP_001091989.1
NM_152759.4 382 Intron NP_689972.3
XM_017019124.1 382 Intron XP_016874613.1
XM_017019125.1 382 Intron XP_016874614.1

View Full Product Details