Product Details

SNP ID
rs79201004
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:122172668 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGGCTGGGCTGTGGATGTTGTTT[G/T]GCGGCTGGGCGTCAGGGCTGAGACA
Phenotype
MIM: 609509
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
IL31 PubMed Links

Gene Details

Gene
IL31
Gene Name
interleukin 31
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001014336.1 266 Missense Mutation CAA,CCA Q80P NP_001014358.1
Gene
LRRC43
Gene Name
leucine rich repeat containing 43
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098519.1 266 Intron NP_001091989.1
NM_152759.4 266 Intron NP_689972.3
XM_017019124.1 266 Intron XP_016874613.1
XM_017019125.1 266 Intron XP_016874614.1

View Full Product Details