Product Details

SNP ID
rs80113116
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:6947222 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTACGTGTTGAGCATCTATTATGCA[A/C]CAGGTGCTGTTTAAGATGCTGGTAA
Phenotype
MIM: 607462 MIM: 615140 MIM: 176883
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ATN1 PubMed Links

Gene Details

Gene
ATN1
Gene Name
atrophin 1
There are no transcripts associated with this gene.

Gene
C12orf57
Gene Name
chromosome 12 open reading frame 57
There are no transcripts associated with this gene.

Gene
PTPN6
Gene Name
protein tyrosine phosphatase, non-receptor type 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002831.5 Intron NP_002822.2
NM_080548.4 Intron NP_536858.1
NM_080549.3 Intron NP_536859.1
XM_006718994.1 Intron XP_006719057.1
XM_011520988.1 Intron XP_011519290.1
Gene
RNU7-1
Gene Name
RNA, U7 small nuclear 1
There are no transcripts associated with this gene.

View Full Product Details