Product Details

SNP ID
rs75912815
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:109897404 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCATGCTGGTCGAGCCCTAAGCCC[G/T]AAAGTTCTGCTAGTGGAGCTTATCT
Phenotype
MIM: 612654
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TCHP PubMed Links

Gene Details

Gene
TCHP
Gene Name
trichoplein keratin filament binding
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143852.1 Intron NP_001137324.1
NM_032300.4 Intron NP_115676.1
XM_011538836.2 Intron XP_011537138.1
XM_011538837.2 Intron XP_011537139.1
XM_017020024.1 Intron XP_016875513.1
XM_017020025.1 Intron XP_016875514.1

View Full Product Details