Product Details

SNP ID
rs79780196
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:41987289 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGTAAGGATTTCTCGGAAAGAAT[G/T]GGACAGCCATGACCCTGGGATCACT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PLA2G4E PubMed Links

Gene Details

Gene
PLA2G4E
Gene Name
phospholipase A2 group IVE
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001206670.1 764 Missense Mutation AAT,CAT N640H NP_001193599.1
XM_011521237.2 764 Missense Mutation AAT,CAT N606H XP_011519539.1
XM_011521238.1 764 Missense Mutation AAT,CAT N264H XP_011519540.1
XM_011521239.1 764 Missense Mutation AAT,CAT N225H XP_011519541.1
Gene
PLA2G4E-AS1
Gene Name
PLA2G4E antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details