Product Details

SNP ID
rs80067811
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:43201951 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGGCTGACTGAGGCTGTGAGGGG[G/T]TGATACTGCTCTGCTTTCTCTGGCA
Phenotype
MIM: 607089 MIM: 177070
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CCNDBP1 PubMed Links

Gene Details

Gene
CCNDBP1
Gene Name
cyclin D1 binding protein 1
There are no transcripts associated with this gene.

Gene
EPB42
Gene Name
erythrocyte membrane protein band 4.2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000119.2 1838 Missense Mutation CAA,CAC Q632H NP_000110.2
NM_001114134.1 1838 Missense Mutation CAA,CAC Q602H NP_001107606.1
XM_005254225.1 1838 Missense Mutation CAA,CAC Q567H XP_005254282.1
XM_011521349.2 1838 Missense Mutation CAA,CAC Q632H XP_011519651.1
XM_011521350.2 1838 Missense Mutation CAA,CAC Q632H XP_011519652.1
XM_011521351.2 1838 Missense Mutation CAA,CAC Q632H XP_011519653.1
XM_011521352.2 1838 Missense Mutation CAA,CAC Q620H XP_011519654.1
XM_011521353.2 1838 Missense Mutation CAA,CAC Q597H XP_011519655.1
XM_011521354.2 1838 Missense Mutation CAA,CAC Q447H XP_011519656.1

View Full Product Details