Product Details
- SNP ID
-
rs79668152
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:49860371 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGATAAGCTCTCCATAGCCTTCTTG[C/G]TGAGCAAAAGCATATCCAGACCTTC
- Phenotype
-
MIM: 609123
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ATP8B4
PubMed Links
Gene Details
- Gene
- ATP8B4
- Gene Name
- ATPase phospholipid transporting 8B4 (putative)
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_024837.3 |
3755 |
Missense Mutation |
CAC,CAG |
H1134Q |
NP_079113.2 |
XM_011522046.2 |
3755 |
Missense Mutation |
CAC,CAG |
H1199Q |
XP_011520348.1 |
XM_011522047.2 |
3755 |
Missense Mutation |
CAC,CAG |
H1162Q |
XP_011520349.1 |
XM_011522048.1 |
3755 |
Missense Mutation |
CAC,CAG |
H1162Q |
XP_011520350.1 |
XM_011522049.2 |
3755 |
Missense Mutation |
CAC,CAG |
H1162Q |
XP_011520351.1 |
XM_011522051.2 |
3755 |
Missense Mutation |
CAC,CAG |
H1162Q |
XP_011520353.1 |
XM_011522052.2 |
3755 |
Missense Mutation |
CAC,CAG |
H1162Q |
XP_011520354.1 |
XM_011522053.1 |
3755 |
Missense Mutation |
CAC,CAG |
H1162Q |
XP_011520355.1 |
XM_011522056.2 |
3755 |
Missense Mutation |
CAC,CAG |
H1264Q |
XP_011520358.2 |
XM_011522058.2 |
3755 |
Missense Mutation |
CAC,CAG |
H1053Q |
XP_011520360.1 |
XM_011522059.1 |
3755 |
Missense Mutation |
CAC,CAG |
H1047Q |
XP_011520361.1 |
XM_011522060.1 |
3755 |
Missense Mutation |
CAC,CAG |
H1035Q |
XP_011520362.1 |
XM_011522061.1 |
3755 |
Missense Mutation |
CAC,CAG |
H1035Q |
XP_011520363.1 |
XM_011522062.1 |
3755 |
Missense Mutation |
CAC,CAG |
H1035Q |
XP_011520364.1 |
XM_011522063.1 |
3755 |
Missense Mutation |
CAC,CAG |
H1035Q |
XP_011520365.1 |
XM_011522064.1 |
3755 |
Missense Mutation |
CAC,CAG |
H950Q |
XP_011520366.1 |
XM_011522069.2 |
3755 |
Missense Mutation |
CAC,CAG |
H879Q |
XP_011520371.1 |
XM_011522070.1 |
3755 |
Missense Mutation |
CAC,CAG |
H681Q |
XP_011520372.1 |
XM_017022587.1 |
3755 |
Missense Mutation |
CAC,CAG |
H1236Q |
XP_016878076.1 |
XM_017022588.1 |
3755 |
Missense Mutation |
CAC,CAG |
H1211Q |
XP_016878077.1 |
XM_017022589.1 |
3755 |
Missense Mutation |
CAC,CAG |
H1201Q |
XP_016878078.1 |
XM_017022590.1 |
3755 |
Missense Mutation |
CAC,CAG |
H1162Q |
XP_016878079.1 |
XM_017022591.1 |
3755 |
Missense Mutation |
CAC,CAG |
H1162Q |
XP_016878080.1 |
XM_017022592.1 |
3755 |
Missense Mutation |
CAC,CAG |
H1134Q |
XP_016878081.1 |
XM_017022593.1 |
3755 |
Intron |
|
|
XP_016878082.1 |
XM_017022594.1 |
3755 |
Missense Mutation |
CAC,CAG |
H1053Q |
XP_016878083.1 |
XM_017022595.1 |
3755 |
Missense Mutation |
CAC,CAG |
H1035Q |
XP_016878084.1 |
XM_017022596.1 |
3755 |
Missense Mutation |
CAC,CAG |
H923Q |
XP_016878085.1 |
XM_017022597.1 |
3755 |
Intron |
|
|
XP_016878086.1 |
XM_017022598.1 |
3755 |
Intron |
|
|
XP_016878087.1 |
XM_017022599.1 |
3755 |
Intron |
|
|
XP_016878088.1 |
View Full Product Details