Product Details

SNP ID
rs77014779
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:55205375 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAATTTTAGAACCGGATGCTTTATT[C/T]GTAGGTCTAATGGGGATGGTGAGAA
Phenotype
MIM: 603868 MIM: 613262
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RAB27A PubMed Links

Gene Details

Gene
RAB27A
Gene Name
RAB27A, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004580.4 1254 UTR 3 NP_004571.2
NM_183234.2 1254 UTR 3 NP_899057.1
NM_183235.2 1254 UTR 3 NP_899058.1
NM_183236.2 1254 UTR 3 NP_899059.1
XM_005254576.4 1254 UTR 3 XP_005254633.1
XM_011521852.1 1254 UTR 3 XP_011520154.1
XM_011521854.1 1254 UTR 3 XP_011520156.1
XM_011521855.2 1254 UTR 3 XP_011520157.1
XM_011521856.2 1254 UTR 3 XP_011520158.1
Gene
RSL24D1
Gene Name
ribosomal L24 domain containing 1
There are no transcripts associated with this gene.

View Full Product Details