Product Details

SNP ID
rs75461420
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:60488841 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTTGTGAAATTTGTGTGCTTAATC[A/C]TCAAGACCTACACACAATTAGAATC
Phenotype
MIM: 610835 MIM: 600825
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ICE2 PubMed Links

Gene Details

Gene
ICE2
Gene Name
interactor of little elongation complex ELL subunit 2
There are no transcripts associated with this gene.

Gene
RORA
Gene Name
RAR related orphan receptor A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002943.3 10183 UTR 3 NP_002934.1
NM_134260.2 10183 UTR 3 NP_599022.1
NM_134261.2 10183 UTR 3 NP_599023.1
NM_134262.2 10183 UTR 3 NP_599024.1
XM_005254584.4 10183 UTR 3 XP_005254641.1
XM_011521874.1 10183 UTR 3 XP_011520176.1
XM_011521875.1 10183 UTR 3 XP_011520177.1
XM_011521877.2 10183 UTR 3 XP_011520179.1
XM_011521878.1 10183 UTR 3 XP_011520180.1
XM_011521879.2 10183 UTR 3 XP_011520181.1
XM_017022466.1 10183 UTR 3 XP_016877955.1
XM_017022467.1 10183 UTR 3 XP_016877956.1
Gene
RORA-AS1
Gene Name
RORA antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details