Product Details

SNP ID
rs76895279
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:90005032 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATGCTTTCTTCATACTTTGACTTG[C/T]ATGTGATTTATTTAGGATTGGTCAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIR3174 PubMed Links

Gene Details

Gene
MIR3174
Gene Name
microRNA 3174
There are no transcripts associated with this gene.

Gene
ZNF710
Gene Name
zinc finger protein 710
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198526.3 Intron NP_940928.2
XM_005254906.3 Intron XP_005254963.1
XM_005254907.3 Intron XP_005254964.1
XM_011521540.2 Intron XP_011519842.1
XM_011521541.2 Intron XP_011519843.1
XM_011521542.2 Intron XP_011519844.1
XM_011521543.2 Intron XP_011519845.1
XM_011521544.1 Intron XP_011519846.1

View Full Product Details