Product Details

SNP ID
rs80287909
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:1810164 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGACAGCGACATCACGTCTCAAAA[A/G]ATGCTTCAGAACACCTCCTGGTAAC
Phenotype
MIM: 138760
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HAGH PubMed Links

Gene Details

Gene
HAGH
Gene Name
hydroxyacylglutathione hydrolase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040427.1 Intron NP_001035517.1
NM_001286249.1 Intron NP_001273178.1
NM_005326.4 Intron NP_005317.2
XM_011522469.2 Intron XP_011520771.1
XM_011522470.2 Intron XP_011520772.1
XM_017023195.1 Intron XP_016878684.1

View Full Product Details