Product Details

SNP ID
rs80312616
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:23296447 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCGGTCCCCTACCCACCCCACTC[A/G]AGGGCTGTCAGTTTGCAACTGTGCA
Phenotype
MIM: 600760
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SCNN1B PubMed Links

Gene Details

Gene
SCNN1B
Gene Name
sodium channel epithelial 1 beta subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000336.2 Intron NP_000327.2
XM_011545913.2 Intron XP_011544215.1
XM_011545914.1 Intron XP_011544216.1
XM_017023525.1 Intron XP_016879014.1
XM_017023526.1 Intron XP_016879015.1

View Full Product Details