Product Details
- SNP ID
-
rs80346735
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:31436377 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGGAAGGACGCGCTGGGAGAGGGAG[G/T]TCTTCTCATTGACACTGTCACCGCA
- Phenotype
-
MIM: 602009
MIM: 602453
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
COX6A2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs74015528] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- COX6A2
- Gene Name
- cytochrome c oxidase subunit 6A2
There are no transcripts associated with this gene.
- Gene
- ITGAD
- Gene Name
- integrin subunit alpha D
There are no transcripts associated with this gene.
- Gene
- ZNF843
- Gene Name
- zinc finger protein 843
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001136509.1 |
1200 |
Missense Mutation |
AAC,ACC |
N158T |
NP_001129981.1 |
View Full Product Details