Product Details

SNP ID
rs78163052
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:3402133 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCAAAAAAACTGCCCAGATCCTGA[G/T]CTCTGCCGCCAGAGCTTCAGACGCT
Phenotype
MIM: 603900
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ZNF174 PubMed Links

Gene Details

Gene
ZNF174
Gene Name
zinc finger protein 174
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001032292.2 944 Missense Mutation GAG,GAT E43D NP_001027463.1
NM_003450.2 944 Missense Mutation GAG,GAT E43D NP_003441.1
Gene
ZSCAN32
Gene Name
zinc finger and SCAN domain containing 32
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001284527.1 944 Intron NP_001271456.1
NM_001284528.1 944 Intron NP_001271457.1
NM_001284529.1 944 Intron NP_001271458.1
NM_001324340.1 944 Intron NP_001311269.1
NM_001324341.1 944 Intron NP_001311270.1
NM_001324342.1 944 Intron NP_001311271.1
NM_001324343.1 944 Intron NP_001311272.1
NM_001324344.1 944 Intron NP_001311273.1
NM_001324345.1 944 Intron NP_001311274.1
NM_001324346.1 944 Intron NP_001311275.1
NM_017810.3 944 Intron NP_060280.2

View Full Product Details