Product Details

SNP ID
rs76955156
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:56363944 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTGCGAGCTTGCTGGAGGAGTTC[A/G]TCCTTACGCTGCACCAGCATGCGCT
Phenotype
MIM: 603243 MIM: 139311
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AMFR PubMed Links

Gene Details

Gene
AMFR
Gene Name
autocrine motility factor receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144.5 1591 Silent Mutation NP_001135.3
NM_001323511.1 1591 Silent Mutation NP_001310440.1
NM_001323512.1 1591 Silent Mutation NP_001310441.1
XM_005255890.3 1591 Silent Mutation XP_005255947.1
Gene
GNAO1
Gene Name
G protein subunit alpha o1
There are no transcripts associated with this gene.

View Full Product Details