Product Details

SNP ID
rs75427202
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:80981478 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCCCTGAGAGTTTTAGTTACCTA[C/T]CAATCAGCTAATAGGTGACTGATAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CMC2 PubMed Links

Gene Details

Gene
CMC2
Gene Name
C-X9-C motif containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020188.3 Intron NP_064573.1
XM_017023461.1 Intron XP_016878950.1
XM_017023462.1 Intron XP_016878951.1
XM_017023463.1 Intron XP_016878952.1
XM_017023464.1 Intron XP_016878953.1
XM_017023465.1 Intron XP_016878954.1
XM_017023466.1 Intron XP_016878955.1
XM_017023467.1 Intron XP_016878956.1
XM_017023468.1 Intron XP_016878957.1
XM_017023469.1 Intron XP_016878958.1
XM_017023470.1 Intron XP_016878959.1

View Full Product Details