Product Details

SNP ID
rs76186241
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:2323097 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTTTAGACATGCAGGCAATGTTG[A/T]GGTTTGTTGTTAAGATGTCTTAATA
Phenotype
MIM: 606477 MIM: 611214
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
SNORD91A PubMed Links

Gene Details

Gene
SNORD91A
Gene Name
small nucleolar RNA, C/D box 91A
There are no transcripts associated with this gene.

Gene
SNORD91B
Gene Name
small nucleolar RNA, C/D box 91B
There are no transcripts associated with this gene.

Gene
SRR
Gene Name
serine racemase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304803.1 4471 Intron NP_001291732.1
NM_021947.2 4471 Intron NP_068766.1
XM_006721565.3 4471 Intron XP_006721628.1
XM_006721566.3 4471 Intron XP_006721629.1
XM_011523974.2 4471 Intron XP_011522276.1
Gene
TSR1
Gene Name
TSR1, ribosome maturation factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018128.4 4471 UTR 3 NP_060598.3

View Full Product Details