Product Details

SNP ID
rs75687931
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:7352562 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGTGTCGCTGACCACCCCAAATC[A/G]GATACGTCCAGACCTCAAGCTCCCT
Phenotype
MIM: 607763 MIM: 609848
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ACAP1 PubMed Links

Gene Details

Gene
ACAP1
Gene Name
ArfGAP with coiled-coil, ankyrin repeat and PH domains 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014716.3 674 Intron NP_055531.1
Gene
KCTD11
Gene Name
potassium channel tetramerization domain containing 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002914.2 674 UTR 5 NP_001002914.1
Gene
TMEM95
Gene Name
transmembrane protein 95
There are no transcripts associated with this gene.

View Full Product Details