Product Details

SNP ID
rs75616699
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.17:1280008 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGCAAGGCCCAGGCCCCCATGGCC[C/T]ACCCGGTGCAGTCCGAGTTTCCTTC
Phenotype
MIM: 615416 MIM: 612211
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
BHLHA9 PubMed Links

Gene Details

Gene
BHLHA9
Gene Name
basic helix-loop-helix family member a9
There are no transcripts associated with this gene.

Gene
TUSC5
Gene Name
tumor suppressor candidate 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_172367.2 346 Missense Mutation CAC,TAC H3Y NP_758955.2

View Full Product Details