Product Details

SNP ID
rs77546775
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:38364785 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGTCGCTTGTGGATGTGGACATTT[C/T]TCAGCGGGGCCTGACCTCTCCACAC
Phenotype
MIM: 608788
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SOCS7 PubMed Links

Gene Details

Gene
SOCS7
Gene Name
suppressor of cytokine signaling 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014598.3 1008 Missense Mutation TCT,TTT S296F NP_055413.1
XM_017024551.1 1008 Missense Mutation TCT,TTT S360F XP_016880040.1
XM_017024552.1 1008 Intron XP_016880041.1

View Full Product Details