Product Details

SNP ID
rs80107361
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:39063520 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACAGTCATGTCTCAGCGAAGAAGT[C/T]GGAGTTCAGCAGCCATCAGAACCAA
Phenotype
MIM: 606826
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC100131347 PubMed Links

Gene Details

Gene
LOC100131347
Gene Name
RAD52 motif containing 1 pseudogene
There are no transcripts associated with this gene.

Gene
PLXDC1
Gene Name
plexin domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020405.4 6035 UTR 3 NP_065138.2

View Full Product Details