Product Details

SNP ID
rs76516244
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:74782567 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTTCCCACCCTTGTACACATGAG[C/T]ATGAAAGTACCAGCAAAAGTAAACA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NAT9 PubMed Links

Gene Details

Gene
NAT9
Gene Name
N-acetyltransferase 9 (putative)
There are no transcripts associated with this gene.

Gene
TMEM104
Gene Name
transmembrane protein 104
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321264.1 Intron NP_001308193.1
NM_017728.3 Intron NP_060198.3
XM_006721963.1 Intron XP_006722026.1
XM_017024797.1 Intron XP_016880286.1
XM_017024798.1 Intron XP_016880287.1
XM_017024799.1 Intron XP_016880288.1

View Full Product Details