Product Details

SNP ID
rs80145926
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:30563283 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGCGCTGACAGCCTCAGAGAGCA[A/G]CAGAGGCCCCTCACTCCTGCAGACT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TBC1D29 PubMed Links

Gene Details

Gene
TBC1D29
Gene Name
TBC1 domain family member 29
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015594.2 460 Missense Mutation AAC,AGC N104S NP_056409.1
XM_011524596.1 460 Missense Mutation AAC,AGC N104S XP_011522898.1

View Full Product Details