Product Details

SNP ID
rs76575208
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:31129994 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGTACAAAAGTGTAAAAGAGAAAT[A/G]AACTTAATCACAAAACTGAAGATTG
Phenotype
MIM: 125643
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DSC1 PubMed Links

Gene Details

Gene
DSC1
Gene Name
desmocollin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004948.3 3467 UTR 3 NP_004939.1
NM_024421.2 3467 UTR 3 NP_077739.1
Gene
DSCAS
Gene Name
DSC1/DSC2 antisense RNA
There are no transcripts associated with this gene.

View Full Product Details