Product Details

SNP ID
rs74711507
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:50974280 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGATATGTGTGGAATACTATGAGA[A/G]ACCAAGAATCCAGACTGTTCTAAAT
Phenotype
MIM: 608079
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ELAC1 PubMed Links

Gene Details

Gene
ELAC1
Gene Name
elaC ribonuclease Z 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018696.2 Intron NP_061166.1

View Full Product Details