Product Details

SNP ID
rs78112898
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:44062222 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAAGCCATAGGGGTTTGATTTCAG[A/G]ACCCCACATGGATACCGAAAGCCAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF223 PubMed Links

Gene Details

Gene
ZNF223
Gene Name
zinc finger protein 223
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013361.5 363 Intron NP_037493.3
XM_017027258.1 363 Intron XP_016882747.1
XM_017027259.1 363 Intron XP_016882748.1
XM_017027260.1 363 UTR 5 XP_016882749.1
Gene
ZNF284
Gene Name
zinc finger protein 284
There are no transcripts associated with this gene.

View Full Product Details