Product Details

SNP ID
rs78968915
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:44100200 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGTAATGCATTCAGGCCAACTTGT[C/T]CAAGGTCACATGACTGGGAAGGGAC
Phenotype
MIM: 194555
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC100379224 PubMed Links

Gene Details

Gene
LOC100379224
Gene Name
uncharacterized LOC100379224
There are no transcripts associated with this gene.

Gene
ZNF224
Gene Name
zinc finger protein 224
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321645.1 Intron NP_001308574.1
NM_013398.3 Intron NP_037530.2
XM_017027261.1 Intron XP_016882750.1

View Full Product Details