Product Details

SNP ID
rs74863643
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:49819225 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTTCAACACAGTGGACTGCGCTCC[C/G]GAGTCCTACGTACAATGTCACGCTC
Phenotype
MIM: 610622 MIM: 610197
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FUZ PubMed Links

Gene Details

Gene
FUZ
Gene Name
fuzzy planar cell polarity protein
There are no transcripts associated with this gene.

Gene
MED25
Gene Name
mediator complex subunit 25
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030973.3 297 Silent Mutation CCC,CCG P78P NP_112235.2
XM_011527353.1 297 Silent Mutation CCC,CCG P78P XP_011525655.1

View Full Product Details