Product Details

SNP ID
rs76493296
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:51125059 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAACTGCTGACGATGCAGAGTTCC[C/G]TGACGGTGCAGGAAGGCCTGTGTGT
Phenotype
MIM: 605640
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SIGLEC9 PubMed Links

Gene Details

Gene
SIGLEC9
Gene Name
sialic acid binding Ig like lectin 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198558.1 152 Missense Mutation CTG,GTG L29V NP_001185487.1
NM_014441.2 152 Missense Mutation CTG,GTG L29V NP_055256.1
XM_006723146.3 152 Missense Mutation CTG,GTG L29V XP_006723209.1
XM_011526730.2 152 Missense Mutation CTG,GTG L29V XP_011525032.1
XM_011526732.2 152 Missense Mutation CTG,GTG L29V XP_011525034.1
XM_017026595.1 152 Missense Mutation CTG,GTG L29V XP_016882084.1
XM_017026596.1 152 Missense Mutation CTG,GTG L29V XP_016882085.1

View Full Product Details