Product Details

SNP ID
rs80323556
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:54773630 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGACCCTCTGGACATCGTGATCA[C/T]AGGTGAGAGTGTCCAGACTTTCTTC
Phenotype
MIM: 604936
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KIR2DL1 PubMed Links

Gene Details

Gene
KIR2DL1
Gene Name
killer cell immunoglobulin like receptor, two Ig domains and long cytoplasmic tail 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014218.2 1011 Missense Mutation ACA,ATA T123I NP_055033.2
XM_011526939.2 1011 Intron XP_011525241.1
XM_017026782.1 1011 Missense Mutation ACA,ATA T70I XP_016882271.1
XM_017026783.1 1011 Missense Mutation ACA,ATA T123I XP_016882272.1
Gene
LOC101928804
Gene Name
uncharacterized LOC101928804
There are no transcripts associated with this gene.

View Full Product Details