Product Details

SNP ID
rs76808668
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:36514396 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTGCTTCTGCCTGAAGATTGTTCC[A/G]CATTCATTACATTTGTAGGGTTTCT
Phenotype
MIM: 613749
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF260 PubMed Links

Gene Details

Gene
ZNF260
Gene Name
zinc finger protein 260
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012756.2 1759 Silent Mutation TGC,TGT C281C NP_001012774.1
NM_001166036.1 1759 Silent Mutation TGC,TGT C281C NP_001159508.1
NM_001166037.1 1759 Silent Mutation TGC,TGT C281C NP_001159509.1
NM_001166038.1 1759 Silent Mutation TGC,TGT C281C NP_001159510.1
XM_011526897.2 1759 Silent Mutation TGC,TGT C281C XP_011525199.1
XM_017026741.1 1759 Silent Mutation TGC,TGT C281C XP_016882230.1

View Full Product Details