Product Details

SNP ID
rs74359855
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:4816604 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGTCCCAAATGACATGTGGCTCC[C/T]AAAAGCCACCTGGAGCTGCTCCATC
Phenotype
MIM: 607601
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TICAM1 PubMed Links

Gene Details

Gene
TICAM1
Gene Name
toll like receptor adaptor molecule 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182919.3 2042 Missense Mutation AGG,GGG R592G NP_891549.1

View Full Product Details