Product Details

SNP ID
rs79572756
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:5920082 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATATTTATGAAAAGGCCAGAAGA[A/C]ACTTTGATTTGGGAAGATAAAACAA
Phenotype
MIM: 114212 MIM: 603327
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CAPS PubMed Links

Gene Details

Gene
CAPS
Gene Name
calcyphosine
There are no transcripts associated with this gene.

Gene
RANBP3
Gene Name
RAN binding protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300865.1 Intron NP_001287794.1
NM_003624.2 Intron NP_003615.2
NM_007320.2 Intron NP_015559.2
NM_007322.2 Intron NP_015561.1
XM_006722928.1 Intron XP_006722991.1
XM_006722929.1 Intron XP_006722992.1
XM_011528391.1 Intron XP_011526693.1
XM_011528392.1 Intron XP_011526694.1
XM_011528393.1 Intron XP_011526695.1
XM_011528394.1 Intron XP_011526696.1
XM_017027405.1 Intron XP_016882894.1
XM_017027406.1 Intron XP_016882895.1
Gene
VMAC
Gene Name
vimentin-type intermediate filament associated coiled-coil protein
There are no transcripts associated with this gene.

View Full Product Details