Product Details

SNP ID
rs79067653
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:9838568 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCTGCTCGCACCTGCTGGTGAAGC[C/A]CAGCCAGTCACGGCGGCCCTCGTCC
Phenotype
MIM: 601052 MIM: 606849
Polymorphism
C/A, Transversion substitution
Allele Nomenclature
Literature Links
PIN1 PubMed Links

Gene Details

Gene
PIN1
Gene Name
peptidylprolyl cis/trans isomerase, NIMA-interacting 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006221.3 Intron NP_006212.1
XM_011528068.2 Intron XP_011526370.1
Gene
UBL5
Gene Name
ubiquitin like 5
There are no transcripts associated with this gene.

View Full Product Details