Product Details

SNP ID
rs76006648
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:13877686 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTACAGCCACACCACCCGAAACTC[A/G]GCAGGCAAGAAGCTGGTCCGGCCTG
Phenotype
MIM: 612746 MIM: 608229
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C19orf57 PubMed Links

Gene Details

Gene
C19orf57
Gene Name
chromosome 19 open reading frame 57
There are no transcripts associated with this gene.

Gene
MIR181C
Gene Name
microRNA 181c
There are no transcripts associated with this gene.

Gene
MIR181D
Gene Name
microRNA 181d
There are no transcripts associated with this gene.

Gene
NANOS3
Gene Name
nanos C2HC-type zinc finger 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098622.2 1304 Silent Mutation TCA,TCG S146S NP_001092092.1
XM_011527964.2 1304 Silent Mutation TCA,TCG S146S XP_011526266.1

View Full Product Details