Product Details

SNP ID
rs74758437
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:17275655 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTGATCCATGCTGAGGGGAATTT[G/T]TGTGTCAGAGAGGGCAAGTGACTTG
Phenotype
MIM: 612766
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
ANKLE1 PubMed Links

Gene Details

Gene
ANKLE1
Gene Name
ankyrin repeat and LEM domain containing 1
There are no transcripts associated with this gene.

Gene
BABAM1
Gene Name
BRISC and BRCA1 A complex member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033549.2 Intron NP_001028721.1
NM_001288756.1 Intron NP_001275685.1
NM_001288757.1 Intron NP_001275686.1
NM_014173.3 Intron NP_054892.2

View Full Product Details