Product Details

SNP ID
rs80113786
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:17460766 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCCCAGCACCAAAGAACAGCAGC[A/C]CCAGCCGGTTCTCCAGCCTGCGACT
Phenotype
MIM: 608791 MIM: 600691
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
NXNL1 PubMed Links

Gene Details

Gene
NXNL1
Gene Name
nucleoredoxin-like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138454.1 151 Missense Mutation GGG,GTG G35V NP_612463.1
Gene
SLC27A1
Gene Name
solute carrier family 27 member 1
There are no transcripts associated with this gene.

View Full Product Details