Product Details

SNP ID
rs76648342
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:51784324 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGGAAACTGGTGTTTGGGAACGC[T/C]GTCAGTAGCACTTGGTTTTTCCACA
Phenotype
MIM: 607343
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
SALL4 PubMed Links

Gene Details

Gene
SALL4
Gene Name
spalt like transcription factor 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318031.1 3394 Missense Mutation AGC,GGC S598G NP_001304960.1
NM_020436.4 3394 Missense Mutation AGC,GGC S1035G NP_065169.1
XM_005260467.3 3394 Intron XP_005260524.1
XM_011528921.2 3394 Missense Mutation AGC,GGC S933G XP_011527223.1
XM_011528922.2 3394 Missense Mutation AGC,GGC S933G XP_011527224.1

View Full Product Details